Merck
Uptake and accumulation of B-group vitamers in Saccharomyces cerevisiae in ethanol-stat fed-batch culture.
World journal of microbiology & biotechnology (2014-05-02)
T Paalme, K Kevvai, A Vilbaste, K Hälvin, I Nisamedtinov
ABSTRACT
The uptake and accumulation of the B-group vitamins thiamine, riboflavin, nicotinamide, pantothenic acid and pyridoxine in Saccharomyces cerevisiae was studied by gradually increasing the specific dosage of vitamins in an ethanol-stat fed-batch culture. Thiamine, nicotinamide, pantothenic acid, and pyridoxine were almost completely taken up at low vitamin dosages. Thiamine was determined to be the major accumulating form of vitamin B1 while most of the assimilated nicotinamide and pantothenic acid accumulated in cofactor forms. Despite the obvious uptake of pyridoxine, accumulation of B6 vitamers was not observed. In contrast with the other vitamins studied, riboflavin began accumulating in the culture medium immediately after vitamin addition was initiated. By the end of the experiment, the apparent uptake of all vitamins exceeded their accumulation in the cells. Variations in the growth rate of yeast at different vitamin dosages demonstrate the importance of balancing the vitamins in the media during cultivation.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Formato di ammonio, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Formato di ammonio, reagent grade, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Formato di ammonio, ≥99.995% trace metals basis
Sigma-Aldrich
Etanolo, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Etanolo, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Etanolo, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Supelco
Etanolo, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Etanolo, standard for GC
Sigma-Aldrich
Etanolo, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Formato di ammonio, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Etanolo, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Etanolo
Sigma-Aldrich
Etanolo, tested according to Ph. Eur.
Sigma-Aldrich
Etanolo, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Fosfatasi, acida, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Etanolo, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
USP
Etanolo, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Etanolo, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)