Merck
  • Phospholipid-modified PEI-based nanocarriers for in vivo siRNA therapeutics against multidrug-resistant tumors.
Phospholipid-modified PEI-based nanocarriers for in vivo siRNA therapeutics against multidrug-resistant tumors.
Gene therapy (2014-10-31)
Sean Essex, Gemma Navarro, Pooja Sabhachandani, Aabha Chordia, Malav Trivedi, Sara Movassaghian, Vladimir P Torchilin
ABSTRACT
Multidrug resistance (MDR) mediated by P-glycoprotein overexpression in solid tumors is a major factor in the failure of many forms of chemotherapy. Here we evaluated phospholipid-modified, low-molecular-weight polyethylenimine (DOPE-PEI) nanocarriers for intravenous delivery of anti-P-pg siRNA to tumors with the final goal of modulating MDR in breast cancer. First, we studied the biodistribution of DOPE-PEI nanocarriers and the effect of PEG coating in a subcutaneous breast tumor model. Four hours postinjection, PEGylated carriers showed an 8% injected dose (ID) accumulation in solid tumor via the enhanced permeability and retention effect and 22% ID in serum due to a prolonged, PEG-mediated circulation. Second, we established the therapeutic efficacy and safety of DOPE-PEI/siRNA-mediated P-gp downregulation in combination with doxorubicin (Dox) chemotherapy in MCF-7/MDR xenografts. Weekly injection of siRNA nanopreparations and Dox for up to 5 weeks sensitized the tumors to otherwise non-effective doses of Dox and decreased the tumor volume by threefold vs controls. This therapeutic improvement in response to Dox was attributed to the significant, sequence-specific P-gp downregulation in excised tumors mediated by the DOPE-PEI formulations.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Colesterolo, Sigma Grade, ≥99%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Colesterolo, powder, BioReagent, suitable for cell culture, ≥99%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Colesterolo, from sheep wool, ≥92.5% (GC), powder
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Estradiol, meets USP testing specifications
Prezzi e disponibilità al momento non sono disponibili
Supelco
Colesterolo, Pharmaceutical Secondary Standard; Certified Reference Material
Prezzi e disponibilità al momento non sono disponibili
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Prezzi e disponibilità al momento non sono disponibili
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Colesterolo, from lanolin, ≥99.0% (GC)
Prezzi e disponibilità al momento non sono disponibili
SAFC
Colesterolo, from sheep wool, Controlled origin, meets USP/NF testing specifications
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Colesterolo, tested according to Ph. Eur.
Prezzi e disponibilità al momento non sono disponibili