Merck
  • Mechanism of N-Acylthiourea-mediated activation of human histone deacetylase 8 (HDAC8) at molecular and cellular levels.
Mechanism of N-Acylthiourea-mediated activation of human histone deacetylase 8 (HDAC8) at molecular and cellular levels.
The Journal of biological chemistry (2015-01-22)
Raushan K Singh, Kyongshin Cho, Satish K R Padi, Junru Yu, Manas Haldar, Tanmay Mandal, Changhui Yan, Gregory Cook, Bin Guo, Sanku Mallik, D K Srivastava
ABSTRACT
We reported previously that an N-acylthiourea derivative (TM-2-51) serves as a potent and isozyme-selective activator for human histone deacetylase 8 (HDAC8). To probe the molecular mechanism of the enzyme activation, we performed a detailed account of the steady-state kinetics, thermodynamics, molecular modeling, and cell biology studies. The steady-state kinetic data revealed that TM-2-51 binds to HDAC8 at two sites in a positive cooperative manner. Isothermal titration calorimetric and molecular modeling data conformed to the two-site binding model of the enzyme-activator complex. We evaluated the efficacy of TM-2-51 on SH-SY5Y and BE(2)-C neuroblastoma cells, wherein the HDAC8 expression has been correlated with cellular malignancy. Whereas TM-2-51 selectively induced cell growth inhibition and apoptosis in SH-SY5Y cells, it showed no such effects in BE(2)-C cells, and this discriminatory feature appears to be encoded in the p53 genotype of the above cells. Our mechanistic and cellular studies on HDAC8 activation have the potential to provide insight into the development of novel anticancer drugs.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Ioduro di propidio, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ioduro di propidio, ≥94% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)