Merck
Electrogenerated chemiluminescence behavior of peptide nanovesicle and its application in sensing dopamine.
Biosensors & bioelectronics (2014-08-19)
Chunxiu Huang, Xu Chen, Yanluo Lu, Hui Yang, Wensheng Yang
ABSTRACT
The electrogenerated chemiluminescence (ECL) behavior of the bioinspired peptide nanovesicles (PNVs) was reported for the first time. The PNVs modified glassy carbon electrodes have shown a stable and efficient cathodic ECL signal with K2S2O8 as coreactant in aqueous solution. The possible ECL reaction mechanism was proposed. Dopamine (DA) was chosen as a model analyte to study the potential of the PNVs in the ECL analytical application. It was found that the ECL intensity of the PNVs was effectively increased by trace amounts of DA. The limit of detection was estimated to be 3.15 pM (S/N=3). These results suggest that the PNVs could be a new class of promising materials for the ECL design and bioassays in the future due to their fascinating features, such as excellent biocompatibility, tunable composition as well as capability of molecular recognition.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
2-Propanolo, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanolo, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanolo, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanolo, meets USP testing specifications
Sigma-Aldrich
2-Propanolo, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
2-Propanolo, anhydrous, 99.5%
Sigma-Aldrich
2-Propanolo, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanolo, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanolo, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanolo, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanolo, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanolo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanolo, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanolo, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Supelco
2-Propanolo, analytical standard
Supelco
1,1,1,3,3,3-esafluoro-2-propanolo, derivatization grade (GC derivatization), LiChropur, ≥99.8%
Sigma-Aldrich
2-Propanolo, suitable for HPLC, 99.5%
USP
2-Propanolo, United States Pharmacopeia (USP) Reference Standard
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Propanolo, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles