Merck
  • Pregnancy induces resistance to the anorectic effect of hypothalamic malonyl-CoA and the thermogenic effect of hypothalamic AMPK inhibition in female rats.
Pregnancy induces resistance to the anorectic effect of hypothalamic malonyl-CoA and the thermogenic effect of hypothalamic AMPK inhibition in female rats.
Endocrinology (2014-12-24)
Pablo B Martínez de Morentin, Ricardo Lage, Ismael González-García, Francisco Ruíz-Pino, Luís Martins, Diana Fernández-Mallo, Rosalía Gallego, Johan Fernø, Rosa Señarís, Asish K Saha, Sulay Tovar, Carlos Diéguez, Rubén Nogueiras, Manuel Tena-Sempere, Miguel López
ABSTRACT
During gestation, hyperphagia is necessary to cope with the metabolic demands of embryonic development. There were three main aims of this study: Firstly, to investigate the effect of pregnancy on hypothalamic fatty acid metabolism, a key pathway for the regulation of energy balance; secondly, to study whether pregnancy induces resistance to the anorectic effect of fatty acid synthase (FAS) inhibition and accumulation of malonyl-coenzyme A (CoA) in the hypothalamus; and, thirdly, to study whether changes in hypothalamic AMPK signaling are associated with brown adipose tissue (BAT) thermogenesis during pregnancy. Our data suggest that in pregnant rats, the hypothalamic fatty acid pathway shows an overall state that should lead to anorexia and elevated BAT thermogenesis: decreased activities of AMP-activated protein kinase (AMPK), FAS, and carnitine palmitoyltransferase 1, coupled with increased acetyl-CoA carboxylase function with subsequent elevation of malonyl-CoA levels. This profile seems dependent of estradiol levels but not prolactin or progesterone. Despite the apparent anorexic and thermogenic signaling in the hypothalamus, pregnant rats remain hyperphagic and display reduced temperature and BAT function. Actually, pregnant rats develop resistance to the anorectic effects of central FAS inhibition, which is associated with a reduction of proopiomelanocortin (POMC) expression and its transcription factors phospho-signal transducer and activator of transcription 3, and phospho-forkhead box O1. This evidence demonstrates that pregnancy induces a state of resistance to the anorectic and thermogenic actions of hypothalamic cellular signals of energy surplus, which, in parallel to the already known refractoriness to leptin effects, likely contributes to gestational hyperphagia and adiposity.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Dimetil solfossido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetil solfossido, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimetil solfossido, Molecular Biology
Sigma-Aldrich
Dimetil solfossido, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimetil solfossido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetil solfossido, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimetil solfossido, anhydrous, ≥99.9%
Sigma-Aldrich
Dimetil solfossido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimetil solfossido, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimetil solfossido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimetil solfossido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, ≥95% (HPLC)
Sigma-Aldrich
Dimetil solfossido, PCR Reagent
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, ≥99%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, Grade AA-1
Sigma-Aldrich
Dimetil solfossido, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Estradiol 3-benzoate, ≥97%
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material