Merck
  • Atypical ubiquitin E3 ligase complex Skp1-Pam-Fbxo45 controls the core epithelial-to-mesenchymal transition-inducing transcription factors.
Atypical ubiquitin E3 ligase complex Skp1-Pam-Fbxo45 controls the core epithelial-to-mesenchymal transition-inducing transcription factors.
Oncotarget (2014-12-03)
Ming Xu, Changhong Zhu, Xian Zhao, Cheng Chen, Hailong Zhang, Haihua Yuan, Rong Deng, Jinzhuo Dou, Yanli Wang, Jian Huang, Qin Chen, Bin Jiang, Jianxiu Yu
ABSTRACT
Epithelial-mesenchymal transition (EMT) plays a critical role in the development of tumor metastases by enhancing migration/invasion. One of the hallmarks of EMT is loss of E-cadherin and gain of N-cadherin expression, which are regulated by the core EMT-inducing transcription factors (EMT-TFs), such as Zeb1/2, Snai1/2 and Twist1. Here, we find that EMT-TFs can be dynamically degraded by an atypical ubiquitin E3 ligase complex Skp1-Pam-Fbxo45 (SPFFbxo45) through the ubiquitin proteasome system (UPS). The key step is recognition of EMT-TFs by Fbxo45 through its SPRY domain for Zeb2, or F-box domain for the other three EMT-TFs Snai1, Snai2 and Twist1, respectively. The K48-linkaged ubiquitination capability on Zeb2 relies on its functional SBD domain. In addition, miR-27a* can directly down-regulate the expression of Fbxo45, preventing degradation of EMT-TFs and thus ensuring EMT phenotype. We suggest that Fbxo45 is a key node of the miR-27a*/Fbxo45/EMT-TFs signaling axis.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo monoclonale ANTI-FLAG® M2, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Sodio cloruro, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodio cloruro, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodio cloruro, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodio cloruro, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodio cloruro, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
SAFC
Sodio cloruro, 5 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodio cloruro, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodio cloruro, BioUltra, Molecular Biology, ~5 M in H2O
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodio cloruro, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Millipore
Protein G Plus/Protein A Agarose Suspension, Protein G PLUS/Protein A-Agarose mixture specifically formulated for immunoprecipitation.
Sigma-Aldrich
Sodio cloruro, 0.85%
Sigma-Aldrich
Sodio cloruro, tested according to Ph. Eur.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone V9, ascites fluid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl