Merck
  • Human placental eXpanded (PLX) mesenchymal-like adherent stromal cells confer neuroprotection to nerve growth factor (NGF)-differentiated PC12 cells exposed to ischemia by secretion of IL-6 and VEGF.
Human placental eXpanded (PLX) mesenchymal-like adherent stromal cells confer neuroprotection to nerve growth factor (NGF)-differentiated PC12 cells exposed to ischemia by secretion of IL-6 and VEGF.
Biochimica et biophysica acta (2014-12-03)
Adi Lahiani, Efrat Zahavi, Nir Netzer, Racheli Ofir, Lena Pinzur, Shani Raveh, Hadar Arien-Zakay, Ephraim Yavin, Philip Lazarovici
ABSTRACT
Mesenchymal stem cells are potent candidates in stroke therapy due to their ability to secrete protective anti-inflammatory cytokines and growth factors. We investigated the neuroprotective effects of human placental mesenchymal-like adherent stromal cells (PLX) using an established ischemic model of nerve growth factor (NGF)-differentiated pheochromocytoma PC12 cells exposed to oxygen and glucose deprivation (OGD) followed by reperfusion. Under optimal conditions, 2 × 10⁵ PLX cells, added in a trans-well system, conferred 30-60% neuroprotection to PC12 cells subjected to ischemic insult. PC12 cell death, measured by LDH release, was reduced by PLX cells or by conditioned medium derived from PLX cells exposed to ischemia, suggesting the active release of factorial components. Since neuroprotection is a prominent function of the cytokine IL-6 and the angiogenic factor VEGF165, we measured their secretion using selective ELISA of the cells under ischemic or normoxic conditions. IL-6 and VEGF165 secretion by co-culture of PC12 and PLX cells was significantly higher under ischemic compared to normoxic conditions. Exogenous supplementation of 10 ng/ml each of IL-6 and VEGF165 to insulted PC12 cells conferred neuroprotection, reminiscent of the neuroprotective effect of PLX cells or their conditioned medium. Growth factors as well as co-culture conditioned medium effects were reduced by 70% and 20% upon pretreatment with 240 ng/ml Semaxanib (anti VEGF165) and/or 400 ng/ml neutralizing anti IL-6 antibody, respectively. Therefore, PLX-induced neuroprotection in ischemic PC12 cells may be partially explained by IL-6 and VEGF165 secretion. These findings may also account for the therapeutic effects seen in clinical trials after treatment with these cells.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Perossido di idrogeno, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Perossido di idrogeno, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Perossido di idrogeno, 50 wt. % in H2O, stabilized
Sigma-Aldrich
2,6-di-tert-butil-4-metilfenolo, ≥99.0% (GC), powder
Sigma-Aldrich
Perossido di idrogeno, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Supelco
2,6-di-tert-butil-4-metilfenolo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethyl phosphate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
2,6-di-tert-butil-4-metilfenolo, ≥99%, FCC, FG
Millipore
Perossido di idrogeno, 3%, suitable for microbiology
Supelco
Perossido di idrogeno, ≥30%, for trace analysis
Sigma-Aldrich
Perossido di idrogeno, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Perossido di idrogeno, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Perossido di idrogeno, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Perossido di idrogeno, purum p.a., ≥35% (RT)
USP
2,6-di-tert-butil-4-metilfenolo, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Perossido di idrogeno, 34.5-36.5%
Sigma-Aldrich
2,6-di-tert-butil-4-metilfenolo, purum, ≥99.0% (GC)
Supelco
Perossido di idrogeno, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
2,6-di-tert-butil-4-metilfenolo, tested according to Ph. Eur.
Sigma-Aldrich
Perossido di idrogeno, tested according to Ph. Eur.