Merck
The prenyltransferase UBIAD1 is the target of geranylgeraniol in degradation of HMG CoA reductase.
eLife (2015-03-06)
Marc M Schumacher, Rania Elsabrouty, Joachim Seemann, Youngah Jo, Russell A DeBose-Boyd
ABSTRACT
Schnyder corneal dystrophy (SCD) is an autosomal dominant disorder in humans characterized by abnormal accumulation of cholesterol in the cornea. SCD-associated mutations have been identified in the gene encoding UBIAD1, a prenyltransferase that synthesizes vitamin K2. Here, we show that sterols stimulate binding of UBIAD1 to the cholesterol biosynthetic enzyme HMG CoA reductase, which is subject to sterol-accelerated, endoplasmic reticulum (ER)-associated degradation augmented by the nonsterol isoprenoid geranylgeraniol through an unknown mechanism. Geranylgeraniol inhibits binding of UBIAD1 to reductase, allowing its degradation and promoting transport of UBIAD1 from the ER to the Golgi. CRISPR-CAS9-mediated knockout of UBIAD1 relieves the geranylgeraniol requirement for reductase degradation. SCD-associated mutations in UBIAD1 block its displacement from reductase in the presence of geranylgeraniol, thereby preventing degradation of reductase. The current results identify UBIAD1 as the elusive target of geranylgeraniol in reductase degradation, the inhibition of which may contribute to accumulation of cholesterol in SCD.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Digitonina, Used as non-ionic detergent
Sigma-Aldrich
Digitonina, ~50% (TLC)
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Farnesol, 95%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Geranylgeraniol, ≥85% (GC)
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
L-arginina, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Supelco
Farnesol, stabilized, analytical standard, mixture of isomers
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesterone, VETRANAL®, analytical standard