Merck
  • Herpes Simplex Virus 1 (HSV-1) ICP22 protein directly interacts with cyclin-dependent kinase (CDK)9 to inhibit RNA polymerase II transcription elongation.
Herpes Simplex Virus 1 (HSV-1) ICP22 protein directly interacts with cyclin-dependent kinase (CDK)9 to inhibit RNA polymerase II transcription elongation.
PloS one (2014-09-19)
Justyna Zaborowska, Sonja Baumli, Clelia Laitem, Dawn O'Reilly, Peter H Thomas, Peter O'Hare, Shona Murphy
ABSTRACT
The Herpes Simplex Virus 1 (HSV-1)-encoded ICP22 protein plays an important role in viral infection and affects expression of host cell genes. ICP22 is known to reduce the global level of serine (Ser)2 phosphorylation of the Tyr1Ser2Pro3Thr4Ser5Pro6Ser7 heptapeptide repeats comprising the carboxy-terminal domain (CTD) of the large subunit of RNA polymerase (pol) II. Accordingly, ICP22 is thought to associate with and inhibit the activity of the positive-transcription elongation factor b (P-TEFb) pol II CTD Ser2 kinase. We show here that ICP22 causes loss of CTD Ser2 phosphorylation from pol II engaged in transcription of protein-coding genes following ectopic expression in HeLa cells and that recombinant ICP22 interacts with the CDK9 subunit of recombinant P-TEFb. ICP22 also interacts with pol II in vitro. Residues 193 to 256 of ICP22 are sufficient for interaction with CDK9 and inhibition of pol II CTD Ser2 phosphorylation but do not interact with pol II. These results indicate that discrete regions of ICP22 interact with either CDK9 or pol II and that ICP22 interacts directly with CDK9 to inhibit expression of host cell genes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Glicerolo, ACS reagent, ≥99.5%
Sigma-Aldrich
Glicerolo, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glicerolo, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Glicerolo, ≥99.5%
Sigma-Aldrich
Glicerolo, 83.5-89.5% (T)
Sigma-Aldrich
Glicerolo, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glicerolo, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glicerolo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glicerolo, BioXtra, ≥99% (GC)
USP
Glicerina, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glicerolo, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glicerolo, FCC, FG
Sigma-Aldrich
Glicerolo, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glicerolo, meets USP testing specifications
Sigma-Aldrich
Glicerolo, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Glicerolo, analytical standard
Sigma-Aldrich
Anticorpo anti-CTCF, serum, Upstate®
Sigma-Aldrich
Glicerolo, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glicerolo, Vetec, reagent grade, 99%