Merck
  • Wogonin induces apoptosis and endoplasmic reticulum stress in HL-60 leukemia cells through inhibition of the PI3K-AKT signaling pathway.
Wogonin induces apoptosis and endoplasmic reticulum stress in HL-60 leukemia cells through inhibition of the PI3K-AKT signaling pathway.
Oncology reports (2015-04-08)
Chengjun Hu, Maozhong Xu, Rujuan Qin, Weifeng Chen, Xin Xu
ABSTRACT
Wogonin is a flavonoid isolated from Scutellaria baicalensis root and has multiple pharmacological effects, including anticancer effects. Recent studies have shown that wogonin induces cell cycle arrest and reverses multi-drug resistance in the human K562 leukemia cell line. However, its pharmacological function in the apoptosis of leukemia cells remains unknown. Therefore, we hypothesized that wogonin can induce apoptosis in the HL-60 leukemia cell line. In the present study, the HL-60 cells were treated with different doses of wogonin (0-150 µM). Wogonin inhibited the viability of HL-60 cells in a dose-dependent and time-dependent manner. Flow cytometry and analyses of caspase and PARP-1 activation and the Bax/Bcl-2 ratio, demonstrated that the cytotoxic effect of wogonin on HL-60 cells was mediated by caspase-dependent and mitochondrial-dependent apoptosis. Wogonin also induced the expression of certain members of the endoplasmic reticulum (ER) stress pathway (CHOP, GRP94 and GRP78) and the activation of multiple branches of ER stress transducers (IRE1α, PERK-eIF2α and ATF6) in the HL-60 cells. In addition, wogonin reduced the phosphorylation of PI3K and AKT in the HL-60 cells. Furthermore, constitutive activation of AKT induced by adenoviral vectors inhibited the pro-apoptotic effects and ER stress induced by wogonin in the HL-60 cells. In summary, our results indicated that wogonin induced apoptosis and ER stress in HL-60 cells, which was mediated by the inhibition of the PI3K-AKT signaling pathway.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Dimetil solfossido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetil solfossido, Molecular Biology
Sigma-Aldrich
Dimetil solfossido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetil solfossido, anhydrous, ≥99.9%
Sigma-Aldrich
Saccarosio, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimetil solfossido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Saccarosio, ≥99.5% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Saccarosio, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccarosio, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Tiazolil blu tetrazolio bromuro, 98%
Sigma-Aldrich
Ioduro di propidio, ≥94.0% (HPLC)
Sigma-Aldrich
Digitonina, Used as non-ionic detergent
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Fenil metansolfonile fluoruro, ≥98.5% (GC)
Sigma-Aldrich
Tiazolil blu tetrazolio bromuro, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimetil solfossido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Digitonina, ~50% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccarosio, ≥99.5% (GC)
Sigma-Aldrich
Saccarosio, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccarosio, ACS reagent
Sigma-Aldrich
Fenil metansolfonile fluoruro, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimetil solfossido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Saccarosio, ≥99.5% (GC), Grade II, suitable for plant cell culture