Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
250 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 250,00 Kč |
1000 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 7 900,00 Kč |
O této položce
2 250,00 Kč
material
glass
sterility
sterile; γ-irradiated
feature
graduations: no
packaging
pack of 25 ea (packed in blisterpacks)
manufacturer/tradename
DWK Life Sciences Sigma-Aldrich
technique(s)
cell culture: suitable
L
5 3/4 in.
1 of 1
Tato položka | |||
|---|---|---|---|
| description non-plugged, L 5 3/4 in. | description cotton plugged, L 5 3/4 in. | description cotton plugged, L 9 in. | description non-plugged, L 9 in. |
| manufacturer/tradename DWK Life Sciences Sigma-Aldrich | manufacturer/tradename DWK Life Sciences Sigma-Aldrich | manufacturer/tradename DWK Life Sciences Sigma-Aldrich | manufacturer/tradename DWK Life Sciences Sigma-Aldrich |
| feature graduations: no | feature plugged, graduations: no | feature graduations: no, plugged | feature graduations: no |
| sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated |
| material glass | material glass | material glass | material glass |
| packaging pack of 25 ea (packed in blisterpacks) | packaging pack of 25 ea (packed in blisterpacks) | packaging pack of 25 ea (acked in blisterpacks) | packaging pack of 25 ea (packed in blisterpacks) |
| L 5 3/4 in. | L 5 3/4 in. | L - | L 9 in. |
Still not finding the right product?
Explore all of our products under Sigma Pasteur pipettes
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?