Merck
  • A combinatorial systematic evolution of ligands by exponential enrichment method for selection of aptamer against protein targets.
A combinatorial systematic evolution of ligands by exponential enrichment method for selection of aptamer against protein targets.
Applied microbiology and biotechnology (2015-08-22)
Bhairab Mondal, Shylaja Ramlal, Padma Sudha Rani Lavu, Harishchandra Sreepathy Murali, Harsh Vardhan Batra
RESUMEN
Aptamers are synthetic DNA recognition elements which form unique conformations that enable them to bind specifically to their targets. In the present study, an attempt was made to standardize a new modified combinatorial method comprising of Ni-NTA affinity Systematic Evolution of Ligands by Exponential Enrichment (SELEX; based on affinity between His tag protein and Ni-NTA), membrane SELEX (based on immobilization of protein on nitrocellulose membrane), and microtiter plate based SELEX (to monitor affinity and to enrich the selected aptamers) for protein targets. For experimental evaluation, staphylococcal interotoxin B was the molecule chosen. The new combinatorial method enhanced selection ability up to 51.20 % in comparison with individual conventional procedures. Employing this method following six rounds of selection, high-affinity aptamers with very different properties could be obtained with a dissociation constant (K d) value as low as 34.72 ± 25.09 nM. The optimal aptamers could be employed in fluorescence binding assay, enzyme-linked oligonucleotide assays, and aptamer-based Western blot assay for characterization and detection. These results pave a potential path without using of any robotics for high-throughput generation of aptamers with advantages in terms of rapidity, simplicity, and ease in handling.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Peróxido de hidrógeno solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hidróxido de sodio, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Fosfato de potasio dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Hidróxido de sodio, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Peróxido de hidrógeno solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hidróxido de sodio solution, 50% in H2O
Sigma-Aldrich
Glicina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Glicina, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Hidróxido de sodio solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Hidróxido de sodio solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Cloruro de calcio solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Hidróxido de sodio, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hidróxido de sodio, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Peróxido de hidrógeno solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Hidróxido de sodio, reagent grade, 97%, powder
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Glicina, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glicina, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Hidróxido de sodio, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Hidróxido de sodio solution, 5.0 M
Sigma-Aldrich
3,3′,5,5′-Tetrametilbencidina, ≥99%
Sigma-Aldrich
Hidróxido de sodio, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)