Merck
Magnetic-nanobead-based competitive enzyme-linked aptamer assay for the analysis of oxytetracycline in food.
Analytical and bioanalytical chemistry (2015-04-10)
Chunxia Lu, Zonggui Tang, Changbin Liu, Lichao Kang, Fengxia Sun
RESUMEN
This study presents a novel analytical method for the detection of oxytetracycline (OTC) in complex food matrices based on a direct competitive enzyme-linked aptamer assay and magnetic separation technology. In this protocol, free OTC competed with horseradish peroxidase labeled OTC (OTC-HRP) for binding to the OTC aptamer immobilized on magnetic beads. The parameters that can affect the response, such as avidin concentration, aptamer concentration, OTC-HRP concentration, incubation temperature, incubation time, blocking agent, and binding buffer, were optimized. Under the optimal conditions, the linear range for the OTC concentration detection is 0.5-100 ng mL(-1), with a concentration of OTC needed to obtain 50 % of the maximum signal of 14.47 ng mL(-1). The limit of detection and the limit of quantitation were 0.88 and 3.40 ng mL(-1), respectively. There was no obvious cross-reactivity with most of the tetracycline pesticides. The recovery rates ranged from 71.0 to 91.2 % for the food samples, including chicken, milk, and honey, and the relative standard deviation was less than 15.0 %. The proposed method was applied to measure OTC in real samples, and was validated using high-performance liquid chromatography. This method has the advantages of magnetic separation and the concentration effect of magnetic nanoparticles, the specificity of the aptamer, and the high-throughput of microtiter plates; it offers a promising approach for the screening of OTC because it is simple, rapid, highly sensitive, and has low cost.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2- Mercaptoetanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2- Mercaptoetanol, ≥99.0%
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Doxorrubicina hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
2- Mercaptoetanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorrubicina hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Cloruro de sodio, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Cloruro de sodio, tablet
Sigma-Aldrich
Cloruro de sodio, random crystals, 99.9% trace metals basis