Merck
Evolution of Robustness to Protein Mistranslation by Accelerated Protein Turnover.
PLoS biology (2015-11-07)
Dorottya Kalapis, Ana R Bezerra, Zoltán Farkas, Peter Horvath, Zoltán Bódi, Andreea Daraba, Béla Szamecz, Ivo Gut, Mónica Bayes, Manuel A S Santos, Csaba Pál
RESUMEN
Translational errors occur at high rates, and they influence organism viability and the onset of genetic diseases. To investigate how organisms mitigate the deleterious effects of protein synthesis errors during evolution, a mutant yeast strain was engineered to translate a codon ambiguously (mistranslation). It thereby overloads the protein quality-control pathways and disrupts cellular protein homeostasis. This strain was used to study the capacity of the yeast genome to compensate the deleterious effects of protein mistranslation. Laboratory evolutionary experiments revealed that fitness loss due to mistranslation can rapidly be mitigated. Genomic analysis demonstrated that adaptation was primarily mediated by large-scale chromosomal duplication and deletion events, suggesting that errors during protein synthesis promote the evolution of genome architecture. By altering the dosages of numerous, functionally related proteins simultaneously, these genetic changes introduced large phenotypic leaps that enabled rapid adaptation to mistranslation. Evolution increased the level of tolerance to mistranslation through acceleration of ubiquitin-proteasome-mediated protein degradation and protein synthesis. As a consequence of rapid elimination of erroneous protein products, evolution reduced the extent of toxic protein aggregation in mistranslating cells. However, there was a strong evolutionary trade-off between adaptation to mistranslation and survival upon starvation: the evolved lines showed fitness defects and impaired capacity to degrade mature ribosomes upon nutrient limitation. Moreover, as a response to an enhanced energy demand of accelerated protein turnover, the evolved lines exhibited increased glucose uptake by selective duplication of hexose transporter genes. We conclude that adjustment of proteome homeostasis to mistranslation evolves rapidly, but this adaptation has several side effects on cellular physiology. Our work also indicates that translational fidelity and the ubiquitin-proteasome system are functionally linked to each other and may, therefore, co-evolve in nature.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Glicerol, ACS reagent, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glicerol, Molecular Biology, ≥99.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloroformo, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glicerol, ReagentPlus®, ≥99.0% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sulfato de magnesio, anhydrous, ReagentPlus®, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloroformo, contains ethanol as stabilizer, ACS reagent, ≥99.8%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, ≥99.0% (GC), dust-free pellets
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, ACS reagent, ≥99.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥98.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, ReagentPlus®, ≥98.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glicerol, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glicerol solution, 83.5-89.5% (T)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloroformo, contains amylenes as stabilizer, ACS reagent, ≥99.8%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sulfato de magnesio, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
2-Nitrofenil β-D-galactopiranósido, ≥98% (enzymatic)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, BioUltra, Molecular Biology, ≥99.0% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glicerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glicerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sulfato de magnesio, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloroformo, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloroformo, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glicerol, BioXtra, ≥99% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Dodecilsulfatosódico, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad