Merck
  • Development of a suspect and non-target screening approach to detect veterinary antibiotic residues in a complex biological matrix using liquid chromatography/high-resolution mass spectrometry.
Development of a suspect and non-target screening approach to detect veterinary antibiotic residues in a complex biological matrix using liquid chromatography/high-resolution mass spectrometry.
Rapid communications in mass spectrometry : RCM (2015-11-14)
Morgan Solliec, Audrey Roy-Lachapelle, Sébastien Sauvé
RESUMEN
Swine manure can contain a wide range of veterinary antibiotics, which could enter the environment via manure spreading on agricultural fields. A suspect and non-target screening method was applied to swine manure samples to attempt to identify veterinary antibiotics and pharmaceutical compounds for a future targeted analysis method. A combination of suspect and non-target screening method was developed to identify various veterinary antibiotic families using liquid chromatography coupled with high-resolution mass spectrometry (LC/HRMS). The sample preparation was based on the physicochemical parameters of antibiotics for the wide scope extraction of polar compounds prior to LC/HRMS analysis. The amount of data produced was processed by applying restrictive thresholds and filters to significantly reduce the number of compounds found and eliminate matrix components. The suspect and non-target screening was applied on swine manure samples and revealed the presence of seven common veterinary antibiotics and some of their relative metabolites, including tetracyclines, β-lactams, sulfonamides and lincosamides. However, one steroid and one analgesic were also identified. The occurrence of the identified compounds was validated by comparing their retention times, isotopic abundance patterns and fragmentation patterns with certified standards. This identification method could be very useful as an initial step to screen for and identify emerging contaminants such as veterinary antibiotics and pharmaceuticals in environmental and biological matrices prior to quantification.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Agua, Nuclease-Free Water, for Molecular Biology
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de sodio dibasic, ACS reagent, ≥99.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido fórmico, reagent grade, ≥95%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Agua, sterile-filtered, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥96%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido cítrico, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de sodio dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Agua, Deionized
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido cítrico, ACS reagent, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de sodio dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥88%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Agua, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Agua, Molecular Biology, sterile filtered
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido cítrico, ≥99.5%, FCC, FG
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido cítrico, 99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de sodio dibasic, ReagentPlus®, ≥99.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de sodio dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Trimetoprim, ≥98.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido cítrico, anhydrous, suitable for cell culture, suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de sodio dibasic, BioXtra, ≥99.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de sodio dibasic, Molecular Biology, ≥98.5% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad