Skip to Content
Merck

ENAH90822AB1

Enamine

methyl 5-(chlorosulfonyl)thieno[3,2-b]thiophene-2-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 121,030.00
250 MG
HUF 172,900.00
500 MG
HUF 272,194.00
1 G
HUF 349,258.00
2.5 G
HUF 684,190.00
5 G
HUF 1,012,206.00
10 G
HUF 1,501,266.00

Vendor SKU: EN300-1588214-100MG

HUF 121,030.00


Ships in 14 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
296.78
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

methyl 5-(chlorosulfonyl)thieno[3,2-b]thiophene-2-carboxylate

Inchi Code

1S/C8H5ClO4S3/c1-13-8(10)6-2-4-5(14-6)3-7(15-4)16(9,11)12/h2-3H,1H3

InChI key

OJXQLPORXIFZMX-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1588214

MSDS

https://www.enaminestore.com/catalog/EN300-1588214


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service