Merck
GC Analysis of Nitrogen and Nitrous Oxide on Carboxen®-1010 PLOT
GC Analysis of Nitrogen and Nitrous Oxide on Carboxen®-1010 PLOT suitable for GC
Materials
related product
Product No.
Description
Pricing
Carboxen®-1010 PLOT Capillary GC Column
L × I.D. 30 m × 0.53 mm, average thickness 30 μm
CONDITIONS
column
Carboxen-1010 PLOT, 30 m x 0.53 mm I.D. (25467)
oven
150 °C
inj. temp.
200 °C
detector
TCD, 230 °C
carrier gas
helium, 3.0 mL/min
injection
10 μL, direct using a sample valve, 150 °C
sample
500 ppm nitrous oxide in nitrogen
Description
Legal Information
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany