Merck
UHPLC-MS/MS Analysis of Multiple Mycotoxins on Titan C18
UHPLC-MS/MS Analysis of Multiple Mycotoxins on Titan C18 application for UHPLC-MS
Materials
related product
Product No.
Description
Pricing
Deoxynivalenol solution
100 μg/mL in acetonitrile, analytical standard
Fumonisin Mixture
~50 μg/mL in acetonitrile: water (each of FB1 and FB2), analytical standard
Fumonisin B1
reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
15-Acetyldeoxynivalenol
reference material
3-Acetyldeoxynivalenol
Citrinin solution
~100 μg/mL in acetonitrile, analytical standard
Aflatoxin Mix
certified reference material, in methanol (varied conc.), ampule of 5 × 1 mL
Ochratoxin A solution
certified reference material, 50 μg/mL in benzene: acetic acid (99:1), ampule of 1 mL
Zearalenone solution
certified reference material, 50 μg/mL in acetonitrile, ampule of 1 mL
CONDITIONS
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
[A] 5mm ammonium formate with 0.1% formic acid in water ; [B] 5mm ammonium formate with 0.1% formic acid in acetonitrile
gradient
30% to 60%B in 3 min, to 100%B in 2 min, hold at 100%B for 2 min, 100%B to 30%B in 0.5 min, hold at 30% for 2 min
flow rate
0.4 mL/min
pressure
8557 psi (590 bar)
column temp.
40 °C
detector
MS/MS, ESI(+), MRM
injection
2 μL
Description
Analysis Note
Titan C18 UHPLC column provided rapid resoltion of mulitple mycotoxins.
Legal Information
null