Merck
  • Melatonin and its metabolites accumulate in the human epidermis in vivo and inhibit proliferation and tyrosinase activity in epidermal melanocytes in vitro.
Melatonin and its metabolites accumulate in the human epidermis in vivo and inhibit proliferation and tyrosinase activity in epidermal melanocytes in vitro.
Molecular and cellular endocrinology (2014-08-30)
Tae-Kang Kim, Zongtao Lin, William J Tidwell, We Li, Andrzej T Slominski
ABSTRACT
Melatonin and its metabolites including 6-hydroxymelatonin (6(OH)M), N(1)-acetyl-N(2)-formyl-5-methoxykynuramine (AFMK) and 5-methoxytryptamine (5MT) are endogenously produced in human epidermis. This production depends on race, gender and age. The highest melatonin levels are in African-Americans. In each racial group they are highest in young African-Americans [30-50 years old (yo)], old Caucasians (60-90 yo) and Caucasian females. AFMK levels are the highest in African-Americans, while 6(OH)M and 5MT levels are similar in all groups. Testing of their phenotypic effects in normal human melanocytes show that melatonin and its metabolites (10(-5) M) inhibit tyrosinase activity and cell growth, and inhibit DNA synthesis in a dose dependent manner with 10(-9) M being the lowest effective concentration. In melanoma cells, they inhibited cell growth but had no effect on melanogenesis, except for 5MT which enhanced L-tyrosine induced melanogenesis. In conclusion, melatonin and its metabolites [6(OH)M, AFMK and 5MT] are produced endogenously in human epidermis and can affect melanocyte and melanoma behavior.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acido acetico, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acido acetico, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acido formico, reagent grade, ≥95%
Sigma-Aldrich
Sodio dodecil solfato, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acido formico, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodio dodecil solfato, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acido acetico, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Sodio dodecil solfato, ACS reagent, ≥99.0%
Sigma-Aldrich
Acido acetico, suitable for HPLC
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Acido tricloroacetico, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodio dodecil solfato, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Acido formico, ACS reagent, ≥88%
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, 20% in H2O
Supelco
Sodio dodecil solfato, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Sodio dodecil solfato, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Acido tricloroacetico, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acido acetico, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Acido acetico, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodio dodecil solfato, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acido tricloroacetico, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%