Merck
Antifilarial effect of ursolic acid from Nyctanthes arbortristis: molecular and biochemical evidences.
Parasitology international (2014-07-11)
Prasanta Saini, Prajna Gayen, Deepak Kumar, Ananya Nayak, Niladri Mukherjee, Suprabhat Mukherjee, Bikas C Pal, Santi P Sinha Babu
ABSTRACT
A bio-assay guided fractionation and purification approach was used to examine in vitro antifilarial activities of the crude methanolic extract of Nyctanthes arbortristis as well as fractions and isolated compound. From ethyl-acetate fraction we isolated and identified a triterpenoid compound which has been characterized as ursolic acid (UA) by HPLC and NMR data. We are reporting for the first time isolation and identification of UA from the leaves of N. arbortristis. The crude extract and UA showed significant micro- as well as macrofilaricidal activities against the oocyte, microfilaria and adult of Setaria cervi (S. cervi) by dye exclusion test and MTT reduction assay. Significant microfilaricidal activity of UA was further proved against mf of W. bancrofti by viability assay. The findings thus provide a new lead for development of a suitable filaricide from natural products. The molecular mechanism of UA was investigated by performing TUNEL, Hoechst staining, Annexin V-Cy3, flow cytometric analysis and DNA fragmentation assay. Differential expressions of pro- and anti-apoptotic genes were observed at the transcription and translational levels in a dose-dependent manner. Depletion in the worm GSH level and elevation in the parasite GST, SOD and super oxide anion indicated the generation of ROS. In this investigation we are reporting for the first time that UA acts its antifilarial effect through induction of apoptosis and by downregulating and altering the level of some key antioxidants like GSH, GST and SOD of S. cervi.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Acido acetico, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acido acetico, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Trypan Blue, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Acido acetico, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acido acetico, suitable for HPLC
Sigma-Aldrich
Metanolo, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Bromuro di etidio, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Metanolo, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanolo, Absolute - Acetone free
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
USP
Metanolo, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Metanolo, BioReagent, ≥99.93%
Supelco
Metanolo, analytical standard
Supelco
Metanolo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acido acetico, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Trypan Blue, 0.4%, suitable for microscopy
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Bromuro di etidio, BioReagent, Molecular Biology, 500 μg/mL in H2O
USP
Acido acetico, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Xantina, ≥99%
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Supelco
Acido acetico, analytical standard
Sigma-Aldrich
Acido acetico, ≥99.5%, FCC, FG