Merck
  • Exploring the Effects of Glycosylation and Etherification of the Side Chains of the Anticancer Drug Mitoxantrone.
Exploring the Effects of Glycosylation and Etherification of the Side Chains of the Anticancer Drug Mitoxantrone.
ChemMedChem (2015-08-04)
Pazit Shaul, Kfir B Steinbuch, Eran Blacher, Reuven Stein, Micha Fridman
ABSTRACT
Herein we report the synthesis and biological evaluation of symmetric and asymmetric analogues of the DNA intercalating drug mitoxantrone (MTX) in which the side chains of the parent drug were modified through glycosylation or methyl etherification. Several analogues with glycosylated side chains exhibited higher DNA affinity than the parent MTX. The most potent in vitro cytotoxicity was observed for MTX analogue 8 (1,4-dimethoxy-5,8-bis[2-(2-methoxyethylamino)ethylamino]anthracene-9,10-dione) with methoxy ether containing side chains. Treatment of melanoma-bearing mice with MTX or analogue 8 decreased the intraperitoneal tumor burden relative to untreated mice; the effect of 8 was less pronounced than that of MTX. In vitro metabolism assays of MTX with rabbit liver S9 fraction gave rise to several metabolites; almost no metabolites were detected for MTX analogue 8. The results presented indicate that derivatization of the MTX side chain primary hydroxy groups may result in a significant improvement in DNA affinity and lower susceptibility to the formation of potentially toxic metabolites.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Dimetil solfossido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetil solfossido, Molecular Biology
Sigma-Aldrich
Dimetil solfossido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetil solfossido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-glutammina, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-glutammina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimetil solfossido, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-glutammina
Sigma-Aldrich
Dimetil solfossido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimetil solfossido, PCR Reagent
Sigma-Aldrich
Metotrexato, meets USP testing specifications
Sigma-Aldrich
L-glutammina, BioUltra, ≥99.5% (NT)
SAFC
Methotrexate
Sigma-Aldrich
L-glutammina, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-glutammina
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimetil solfossido, ≥99.6%, ReagentPlus®