Merck
Prenatal Stress Alters the Development of Socioemotional Behavior and Amygdala Neuron Excitability in Rats.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2015-02-27)
David E Ehrlich, Donald G Rainnie
ABSTRACT
Prenatal stress (PS) is a risk factor for neurodevelopmental disorders with diverse ages of onset and socioemotional symptoms. Some PS-linked disorders involve characteristic social deficits, such as autism spectrum disorders and schizophrenia, but PS also promotes anxiety disorders. We propose the diversity of symptoms following PS arises from perturbations to early brain development. To this end, we characterized the effects of PS on the developmental trajectory of physiology of the amygdala, a late-developing center for socioemotional control. We found that PS dampened socioemotional behavior and reduced amygdala neuron excitability in offspring during infancy (at postnatal days (P)10, 14, 17 and 21), preadolescence (day 28), and adulthood (day 60). PS offspring in infancy produced fewer isolation-induced vocalizations and in adulthood exhibited less anxiety-like behavior and deficits in social interaction. PS neurons had a more hyperpolarized resting membrane potential from infancy to adulthood and produced fewer action potentials. Moreover, adult amygdala neurons from PS animals expressed larger action potential afterhyperpolarizations and H-current relative to controls, further limiting excitability. Our results suggest that PS can suppress socioemotional behavior throughout development and produce age-specific alterations to amygdala physiology.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Bicarbonato di sodio, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodio cloruro, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodio cloruro, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodio cloruro, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodio cloruro, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodio cloruro, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodio cloruro, 5 M
SAFC
Sodio cloruro, 5 M
Sigma-Aldrich
Sodio cloruro, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodio cloruro, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodio cloruro, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodio cloruro, 99.999% trace metals basis
Sigma-Aldrich
Bicarbonato di sodio, BioXtra, 99.5-100.5%
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Sodio cloruro, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodio cloruro, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodio cloruro, tablet
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodio cloruro, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodio cloruro, AnhydroBeads, −10 mesh, 99.999% trace metals basis