Merck
Experience-dependent DNA methylation regulates plasticity in the developing visual cortex.
Nature neuroscience (2015-05-26)
Paola Tognini, Debora Napoli, Jonida Tola, Davide Silingardi, Floriana Della Ragione, Maurizio D'Esposito, Tommaso Pizzorusso
ABSTRACT
DNA methylation is an epigenetic repressor mark for transcription dynamically regulated in neurons. We analyzed visual experience regulation of DNA methylation in mice and its involvement in ocular dominance plasticity of the developing visual cortex. Monocular deprivation modulated the expression of factors controlling DNA methylation and exerted opposite effects on DNA methylation and hydroxymethylation in specific plasticity genes. Inhibition of DNA methyltrasferase (DNMT) blocked molecular and functional effects of monocular deprivation, partially reversing the monocular deprivation transcriptional program.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sodio dodecil solfato, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Cloruro di magnesio, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodio dodecil solfato, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Cloruro di magnesio, ≥98%
Sigma-Aldrich
Dietilpirocarbonato, 96% (NT)
Sigma-Aldrich
Sodio dodecil solfato, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
(2-idrossipropil)-β-ciclodestrina, powder
Sigma-Aldrich
Sodio dodecil solfato, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Fenolo, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Fenolo:cloroformio:alcol isoamilico 25:24:1 saturato con 10 mM Tris, pH 8,0, 1 mM EDTA, Supplied with Equilibration buffer, Molecular Biology
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Fenolo, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, 20% in H2O
Sigma-Aldrich
Cloruro di magnesio, powder, <200 μm
Supelco
Sodio dodecil solfato, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Fenolo, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Fenolo, ≥99%
Sigma-Aldrich
Cloruro di magnesio, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
Fenolo, ≥89.0%
Sigma-Aldrich
Fenolo, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Cloruro di magnesio, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodio dodecil solfato, BioXtra, ≥99.0% (GC)