Merck
  • Loss of AXIN1 drives acquired resistance to WNT pathway blockade in colorectal cancer cells carrying RSPO3 fusions.
Loss of AXIN1 drives acquired resistance to WNT pathway blockade in colorectal cancer cells carrying RSPO3 fusions.
EMBO molecular medicine (2017-01-20)
Gabriele Picco, Consalvo Petti, Alessia Centonze, Erica Torchiaro, Giovanni Crisafulli, Luca Novara, Andrea Acquaviva, Alberto Bardelli, Enzo Medico
ABSTRACT
In colorectal cancer (CRC), WNT pathway activation by genetic rearrangements of RSPO3 is emerging as a promising target. However, its low prevalence severely limits availability of preclinical models for in-depth characterization. Using a pipeline designed to suppress stroma-derived signal, we find that RSPO3 "outlier" expression in CRC samples highlights translocation and fusion transcript expression. Outlier search in 151 CRC cell lines identified VACO6 and SNU1411 cells as carriers of, respectively, a canonical PTPRK(e1)-RSPO3(e2) fusion and a novel PTPRK(e13)-RSPO3(e2) fusion. Both lines displayed marked
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
XAV939, ≥98% (HPLC)