Merck
Ezrin enhances line tension along transcellular tunnel edges via NMIIa driven actomyosin cable formation.
Nature communications (2017-06-24)
Caroline Stefani, David Gonzalez-Rodriguez, Yosuke Senju, Anne Doye, Nadia Efimova, Sébastien Janel, Justine Lipuma, Meng Chen Tsai, Daniel Hamaoui, Madhavi P Maddugoda, Olivier Cochet-Escartin, Coline Prévost, Frank Lafont, Tatyana Svitkina, Pekka Lappalainen, Patricia Bassereau, Emmanuel Lemichez
ABSTRACT
Transendothelial cell macroaperture (TEM) tunnels control endothelium barrier function and are triggered by several toxins from pathogenic bacteria that provoke vascular leakage. Cellular dewetting theory predicted that a line tension of uncharacterized origin works at TEM boundaries to limit their widening. Here, by conducting high-resolution microscopy approaches we unveil the presence of an actomyosin cable encircling TEMs. We develop a theoretical cellular dewetting framework to interpret TEM physical parameters that are quantitatively determined by laser ablation experiments. This establishes the critical role of ezrin and non-muscle myosin II (NMII) in the progressive implementation of line tension. Mechanistically, fluorescence-recovery-after-photobleaching experiments point for the upstream role of ezrin in stabilizing actin filaments at the edges of TEMs, thereby favouring their crosslinking by NMIIa. Collectively, our findings ascribe to ezrin and NMIIa a critical function of enhancing line tension at the cell boundary surrounding the TEMs by promoting the formation of an actomyosin ring.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-actinaβ monoclonale, clone AC-74, ascites fluid
Sigma-Aldrich
Isotiocianato di falloidina-tetrametilrodamina B, sequence from Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Falloidina marcata, fluorescina isotiocianato, sequence Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder