Merck
  • A green capillary zone electrophoresis method for the simultaneous determination of piperacillin, tazobactam and cefepime in pharmaceutical formulations and human plasma.
A green capillary zone electrophoresis method for the simultaneous determination of piperacillin, tazobactam and cefepime in pharmaceutical formulations and human plasma.
Biomedical chromatography : BMC (2015-06-11)
Amirah Al-Attas, Jenny Jeehan Nasr, Nahed El-Enany, Fathalla Belal
RESUMEN
A green, novel, rapid, accurate and reliable capillary zone electrophoresis method was developed and validated for the simultaneous determination of piperacillin, tazobactam and cefepime in pharmaceutical preparations. Separation was carried out using fused silica capillary (50 µm i.d. × 48.6 cm and 40.2 cm detection length) and applied potential of 20 kV (positive polarity) and a running buffer containing 15 m m sodium borate buffer adjusted to pH 9.3 with UV detection at 215 nm. Amoxicillin was used as an internal standard. The method was suitably validated according to International Conference on Harmonization guidelines. The method showed good linearity in the ranges of 10-100, 20-400 and 10-400 µg/mL with limits of quantitation of 1.87, 3.17 and 6.97 µg/mL and limits of detection of 0.56, 0.95 and 2.09 µg/mL for tazobactam, piperacillin and cefepime, respectively. The proposed method was successfully applied for the analysis of these drugs in their synthetic mixtures and co-formulated injection vials. The method was extended to the in vitro determination of the two drugs in spiked human plasma. It is considered a 'green' method as it consumes no organic solvents.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Hidróxido de sodio, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hidróxido de sodio, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Ácido fosfórico, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Hidróxido de sodio solution, 50% in H2O
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ácido fosfórico, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Hidróxido de sodio solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Ácido fosfórico, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
Hidróxido de sodio solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hidróxido de sodio, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hidróxido de sodio, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Hidróxido de sodio, reagent grade, 97%, powder
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ácido fosfórico, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Hidróxido de sodio, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Hidróxido de sodio, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Ácido fosfórico, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%