Skip to Content
Merck

05-826-AF488

Anti-Bcl-2 Antibody, clone AW604, Alexa Fluor 488 Conjugate

Synonym(s):

Apoptosis regulator Bcl-2|Bcl-2

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

clone

monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
05-729MABC573SAB1401027
species reactivity

human

species reactivity

feline, canine, human

species reactivity

mouse

species reactivity

human

biological source

mouse

biological source

mouse

biological source

rat

biological source

mouse

clone

monoclonal

clone

100, monoclonal

clone

1D7.2, monoclonal

clone

polyclonal

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable (Paraffin), western blot: suitable

technique(s)

western blot: suitable

technique(s)

western blot: 1 μg/mL

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

General description

Apoptosis regulator Bcl-2 (UniProt P10415) is encoded by the BCL2 gene (Gene ID 596) in human. Bcl-2 is an anti-apoptotic and the founding member of the Bcl-2 family of proteins that play either a pro- or anti-apoptotic role in cell death signaling. Bcl-2 family members are related by regions of sequence and structural homology. The pro-survival members contain four Bcl-2 homology regions (BH1–BH4), whereas the BH3-only members contain only the BH3 amphipathic helix, which mediates their interaction with the groove of multi-domain BCL-2 family members. Human BCL2 gene mutations are well known causes of many cancers, including melanoma, breast, prostate, chronic lymphocytic leukemia, and lung cancer.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 41-54 (C-GAAPAPGIFSSQPG) of human Bcl-2.

Application

This Anti-Bcl-2 Antibody, clone AW604, Alexa Fluor 488 Conjugate is validated for use in Immunocytochemistry, Flow Cytometry for the detection of Bcl-2.

Flow Cytometry Analysis: 1.0 ug of this antibody from a representative lot detected Bc

Analysis Note

Evaluated by Immunocytochemistry in Jurkat cells.

Immunocytochemistry Analysis: A 1:50 dilution of this antibody detected Bcl-2 in Jurkat cells.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service