Merck
SYNC144250
Synthware™ Graham condenser
joint: ST/NS 24/40, hose size 10 mm
Synonim(y):
Synthware C144250, distillation condenser, Inland Revenue condenser
Informacje o tej pozycji
NACRES:
NB.51
UNSPSC Code:
41121800
eCl@ss:
32150326
Pomoc techniczna
Pozwól nam pomóc
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C144250
hose size
10 mm
General description
Synthware™ Graham condenser features a 250 mm jacketed length for superior cooling efficiency. With standardized 24/40 upper and lower joints, it easily fits various laboratory glassware. Made from durable borosilicate glass.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Explore all of our products under Synthware™ Graham condenser
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)