Merck
1.17653
Wi-Fi Dongle
new
for use with MAS-100® Sirius
Zaloguj się aby wyświetlić ceny organizacyjne i kontraktowe
Pozwól nam pomóc
opakowanie
pkg of 1 unit
producent / nazwa handlowa
(Wi-Fi Dongle Sirius)
dilution
(Application - Air monitoring)
Zastosowanie
pharmaceutical (Cosmetic, Medical Device; Hospital)
kompatybilność
for use with MAS-100® Sirius
Opis ogólny
The Wi-Fi Dongle for MAS-100 Sirius is designed to allow various wireless operations for the air sampler such as:
  • Automated date/time synchronization
  • Automated timer calibration
  • Automated user synchronization
  • Active Directory login
  • Wireless access to the browser-based user interface (e.g. audit trail review)
Remote control
Zastosowanie
The Wi-Fi Dongle can be conveniently placed beneath the button plate of the MAS-100 Sirius. Once installed, the instrument seamlessly integrates into your local Wi-Fi network.
Informacje prawne
MAS-100 is a registered trademark of MBV AG