Przejdź do zawartości
Merck

Przejdź do

VANA-1

Isotopic certified reference material of vanillin

certified reference material

Synonim(y):

Isotopic certified reference material of vanillin

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto

Informacje o tej pozycji

NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


grade

certified reference material

agency

traceable to (National Research Council of Canada (NRC))

manufacturer/tradename

National Research Council of Canada (NRC)

application(s)

food and beverages

format

neat

storage temp.

room temp

General description

NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034.For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis.With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.
VANA-1 is a vanillin isotopic Certified Reference Material (CRM) from the National Research Council of Canada (NRC). A unit of VANA-1 consists of approximately 0.75 g of vanillin. This certified reference material is intended to be used for calibration and quality control of bulk and site-specific carbon isotope delta measurements, traceable to the VPDB-LSVEC scale. For more information, please refer to the Certificate of Analysis or Safety Data Sheet.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
VANB1SENS1SLRS6
description

certified reference material

description

certified reference material

description

certified reference material

description

certified reference material

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

format

neat

format

neat

format

neat

format

matrix material

agency

traceable to (National Research Council of Canada (NRC))

agency

traceable to (National Research Council of Canada (NRC))

agency

-

agency

traceable to (National Research Council of Canada (NRC))

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

-

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)


Still not finding the right product?


Piktogramy

Exclamation mark

Słowo ostrzegawcze

Warning

Kody zagrożeń

Hazard Classifications

Eye Irrit. 2

Klasa składowania

11 - Combustible Solids

Temperatura zapłonu (°F)

319.6 - 321.4 °F - closed cup

Temperatura zapłonu (°C)

159.8 - 160.8 °C - closed cup



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów