Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-P-T3
diam. × L
68 mm × 350 mm
compatibility
overhead stirrer equipment
General description
Application
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description flat paddle with holes, PTFE-coated | description centrifugal, PTFE-coated | description 4-arm, PTFE-coated | description flat blade, stainless steel |
| compatibility overhead stirrer equipment | compatibility overhead stirrer equipment | compatibility overhead stirrer equipment | compatibility overhead stirrer equipment |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| material PTFE-coated | material PTFE-coated | material PTFE-coated | material stainless steel |
| diam. × L 68 mm × 350 mm | diam. × L 85 mm × 350 mm , open | diam. × L 65 mm × 300 mm | diam. × L 68 mm × 400 mm |
| manufacturer/tradename Benchmark Scientific IPS2050-P-T3 | manufacturer/tradename Benchmark Scientific IPS2050-P-T2 | manufacturer/tradename Benchmark Scientific IPS2050-P-T4 | manufacturer/tradename Benchmark Scientific IPS2050-P-S1 |
Still not finding the right product?
Explore all of our products under Benchmark Overhead Stirrer Propeller
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?