Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
polypropylene
description
autoclavable
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 7134-0010
L × W × H
15 in. (381 mm) × 13 3/4 in. (350 mm) × 4 3/8 in. (111 mm)
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description L × W × H 15 in. (381 mm) × 13 3/4 in. (350 mm) × 4 3/8 in. (111 mm) | description L × W × H 14 3/4 in. (375 mm) × 9 1/2 in. (241 mm) × 6 in. (153 mm) | description L × W × H 15 in. (381 mm) × 14 in. (356 mm) × 10 1/2 in. (267 mm) | description bright yellow, low-density polyethylene |
| manufacturer/tradename Nalgene® 7134-0010 | manufacturer/tradename Nalgene® 7130-0010 | manufacturer/tradename Nalgene® 7131-0001 | manufacturer/tradename Nalgene® 6505-0010 |
| material polypropylene | material high-density polyethylene | material high-density polyethylene | material low-density polyethylene |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| L × W × H 15 in. (381 mm) × 13 3/4 in. (350 mm) × 4 3/8 in. (111 mm) | L × W × H 14 3/4 in. (375 mm) × 9 1/2 in. (241 mm) × 6 in. (153 mm) | L × W × H 15 in. (381 mm) × 14 in. (356 mm) × 10 1/2 in. (267 mm) | L × W × H - |
| description autoclavable | description - | description - | description - |
Still not finding the right product?
Explore all of our products under Utility carriers, Nalgene®
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?