Merck
CLS431847
Corning® centrifuge bottle
with standard screw cap, capacity 500 mL, polycarbonate
Synonim(y):
centrifugation bottles, digestion bottles, shaking bottles
Wybierz wielkość
24 EA
2480,00 zł
2480,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Informacje o tej pozycji
NACRES:
NB.11
UNSPSC Code:
41121703
Pozwól nam pomóc
Nazwa produktu
Corning® centrifuge bottle, with standard screw cap, capacity 500 mL, polycarbonate
material
clear bottle
flat bottom tube
polycarbonate
round bottle
wide-mouth bottle
sterility
non-sterile
feature
autoclavable
cap
packaging
pkg of 24 ea
manufacturer/tradename
Corning 431847
parameter
13700 × g max. RCF
capacity
500 mL
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 4
Ta pozycja
CLS431846CLS431845CLS431843
manufacturer/tradename
Corning 431847
manufacturer/tradename
Corning 431846
manufacturer/tradename
Corning 431845
manufacturer/tradename
Corning 431843
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
material
clear bottle, polycarbonate , wide-mouth bottle, flat bottom tube, round bottle
material
clear bottle, flat bottom tube, polycarbonate , round bottle, wide-mouth bottle
material
flat bottom tube, polypropylene , round bottle, translucent bottle, wide-mouth bottle
material
clear bottle, flat bottom tube, polycarbonate , round bottle, wide-mouth bottle
parameter
13700 × g max. RCF
parameter
13700 × g max. RCF
parameter
13700 × g max. RCF
parameter
13500 × g max. RCF
packaging
pkg of 24 ea
packaging
pkg of 24 ea
packaging
pkg of 24 ea
packaging
pkg of 36 ea
capacity
500 mL
capacity
500 mL
capacity
500 mL
capacity
250 mL
Features and Benefits
  • Corning® centrifuge bottles have a specially designed cap for improved grip
  • Includes clear graduations and textured marking spot.
  • Bottles are designed with extra-thick walls for high durability
  • Wide mouth for easier liquid filling
  • Compatible with most centrifuges and do not require special buckets
  • Unique design aid researchers to reduce the number of centrifugation cycles and enhance productivity
Legal Information
Corning is a registered trademark of Corning, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)