Merck
CLS480123
Corning® LSE single block, for Corning LSE single block digital dry baths
single block, 20 x 13 mm tubes, 1/cs
Synonim(y):
equipment replacement accessory
Wybierz wielkość
1 EA
519,35 zł
519,35 zł
Cena katalogowa611,00 złZaoszczędź 15%
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Informacje o tej pozycji
NACRES:
NB.15
UNSPSC Code:
41102404
Pozwól nam pomóc
Nazwa produktu
Corning® LSE single block, for Corning LSE single block digital dry baths, single block, 20 x 13 mm tubes, 1/cs
feature
Holds 20 x 13 mm tubes
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 480123
suitability
suitable for (use with Corning LSE digital dry bath heater, single block)
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 4
Ta pozycja
CLS480122CLS480121CLS480125
feature
Holds 20 x 13 mm tubes
feature
Holds 20 x 12 mm tubes
feature
Holds 20 x 10 mm tubes
feature
Holds 12 x 16 mm tubes
manufacturer/tradename
Corning 480123
manufacturer/tradename
Corning 480122
manufacturer/tradename
Corning 480121
manufacturer/tradename
Corning 480125
packaging
case of 1, pack of 1
packaging
case of 1, pack of 1
packaging
case of 1, pack of 1
packaging
case of 1, pack of 1
suitability
suitable for (use with Corning LSE digital dry bath heater, single block)
suitability
suitable for (use with Corning LSE digital dry bath heater, single block)
suitability
suitable for (use with Corning LSE digital dry bath heater, single block)
suitability
suitable for (use with Corning LSE digital dry bath heater, single block)
General description
Corning LSE single blocks are suitable for the following digital dry baths:
CLS6785SB, CLS6786SB, CLS6787SB
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)