Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
red polycarbonate
feature
Holds 25 x 2 mL tubes
packaging
pk of 8
manufacturer/tradename
Heathrow Scientific HS23325A
technique(s)
cryopreservation: suitable
size
76x76x53 mm
General description
- forward sloped base and high contrast indexing on the transparent lid ensure quick visual orientation
- vend and drainage holes
- stackable
- autoclavable
Features and Benefits
- ideal for small sampling
- friction fit, slide on lid
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Holds 25 x 2 mL tubes, red polycarbonate, pk of 8 | description Holds 100 x 2 mL tubes, blue polycarbonate, pk of 4 | description Holds 81 x 2 mL tubes, assorted colors polycarbonate, pk of 4 | description holds 2" freezer boxes, Holds 16 x boxes |
| feature Holds 25 x 2 mL tubes | feature Holds 100 x 2 mL tubes | feature Holds 81 x 2 mL tubes | feature Holds 16 x boxes |
| manufacturer/tradename Heathrow Scientific HS23325A | manufacturer/tradename Heathrow Scientific HS23300D | manufacturer/tradename Heathrow Scientific HS23381B | manufacturer/tradename Heathrow Scientific HS2864 |
| material red polycarbonate | material blue polycarbonate | material assorted colors polycarbonate | material steel |
| packaging pk of 8 | packaging pk of 4 | packaging pk of 4 | packaging - |
| size 76x76x53 mm | size 133x133x53 mm | size 133x133x53 mm | size 139x571x238 mm |
| technique(s) cryopreservation: suitable | technique(s) cryopreservation: suitable | technique(s) cryopreservation: suitable | technique(s) cryopreservation: suitable |
Still not finding the right product?
Explore all of our products under Arctic Squares® Cryostorage Box
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?