Przejdź do
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
usage
sufficient for 200 colorimetric tests
Quality Segment
detection method
colorimetric
relevant disease(s)
autoimmune diseases; gastrointestinal diseases; metabolic disorders
shipped in
ambient
storage temp.
2-8°C
General description
Application
Biochem/physiol Actions
Features and Benefits
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description sufficient for 200 colorimetric tests | description sufficient for 200 tests | description sufficient for 100 colorimetric tests | description sufficient for 100 colorimetric tests |
| Quality Level 200 | Quality Level 200 | Quality Level - | Quality Level 100 |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
| usage sufficient for 200 colorimetric tests | usage sufficient for 200 tests | usage sufficient for 100 colorimetric tests | usage - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C |
| shipped in ambient | shipped in wet ice | shipped in - | shipped in wet ice |
| relevant disease(s) autoimmune diseases; gastrointestinal diseases; metabolic disorders | relevant disease(s) - | relevant disease(s) gastrointestinal diseases | relevant disease(s) gastrointestinal diseases |
Słowo ostrzegawcze
Danger
Kody zagrożeń
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3
Narządy docelowe
Respiratory system
Klasa składowania
3 - Flammable liquids
Co się dzieje?
WGK 2
Temperatura zapłonu (°F)
102.2 °F - closed cup
Temperatura zapłonu (°C)
39 °C - closed cup
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.




