Przejdź do
MTCA8401SK
MTC™ Bio BenchBin™ starter Set
includes: BenchBin™ & 400 red autoclave/biohazard bags
Synonim(y):
bio waste bin, MTC Bio A8401-SK, laboratory bin, waste canoster, waste receptacle
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.15
UNSPSC Code:
24111500
Manufacturer/tradename:
MTC™ Bio A8401-SK
Packaging:
pack of 1 ea
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio A8401-SK
bag size
8.5 in. (21.6 cm) × 11 in. (28 cm)
capacity
1.5 L , bin
General description
BenchBin™ is a personal benchtop container for biohazard and contaminated waste. Compact size fits perfectly anywhere and safely disposes up to tips, tubes, gloves, etc. Features easy push open/close lid that is perfect for quick disposal of biological waste. The cover is easily removed from the inner basin for quick emptying of contents. Ideal size for use with red imprinted autoclavable biohazard bags. BenchBin Starter kit includes one BenchBin and 400 red biohazard bags.
Features and Benefits
  • Compact size
  • Safely disposes tips, tubes, gloves
  • Easy open lid
  • Research or further manufacturing use only, not for food or drug use.
Legal Information
BenchBin is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
MTCA8403SKMTCA8401BMTCA9004SK
description
includes: BenchBin™ & 400 red autoclave/biohazard bags
description
includes 3L BenchBin™ biohazard bin & 400 red autoclave/biohazard bags
description
capacity 1.5 L
description
includes: metal stand & 400 red autoclave bags 8.5in. x 11in. (21.6 x 27.9cm)
manufacturer/tradename
MTC™ Bio A8401-SK
manufacturer/tradename
MTC™ Bio A8403-SK
manufacturer/tradename
MTC™ Bio A8401B
manufacturer/tradename
MTC™ Bio A9004-SK
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
capacity
1.5 L , bin
capacity
3 L
capacity
1.5 L
capacity
-
bag size
8.5 in. (21.6 cm) × 11 in. (28 cm)
bag size
-
bag size
-
bag size
-
Still not finding the right product?
Explore all of our products under MTC™ Bio BenchBin™ starter Set
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)