Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
polypropylene
feature
50 x 5 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio C2592
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 50 x 5 mL tubes | description 2 tier, to hold, 16 x 5 mL macrotubes, pack of 1 ea | description capacity 5 mL, black, non-sterile, pack of 200 ea (2 x bags 100 ea) | description capacity 5 mL, blue, non-sterile, pack of 200 ea (2 x bags 100 ea) |
| manufacturer/tradename MTC™ Bio C2592 | manufacturer/tradename MTC™ Bio C2590 | manufacturer/tradename MTC™ Bio C2500-OB | manufacturer/tradename MTC™ Bio C2500-B |
| feature 50 x 5 mL tubes | feature to hold 16 x 5 mL macrotubes, 2 tier | feature graduation | feature graduation |
| material polypropylene | material acrylonitrile-butadiene-styrene (ABS) | material black , cap (snap cap), polypropylene , conical bottom | material blue , conical bottom , polypropylene , cap (snap cap) |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 200 ea (2 x bags 100 ea) | packaging pack of 200 ea (2 x bags 100 ea) |
Still not finding the right product?
Explore all of our products under MTC™ Bio MacroTubes® rack
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?