Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
sterility
sterile
packaging
pack of 500 ea (50 x bags 10 ea)
manufacturer/tradename
MTC™ Bio M5431
L × W
38 mm × 140 mm
General description
Features and Benefits
- Spreads cultures and liquids evenly
- Ideal for bacterial CFU counts
- Smooth edges - minimizes gouging
- Pre-sterilized - no flaming required
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description L × W 38 mm × 140 mm, sterile, pack of 500 ea (50 x bags 10 ea) | description L × W 38 mm × 140 mm, sterile, pack of 500 ea (individually wrapped) | description W × L 38 mm × 156 mm, pack of 500 ea (50 x bags 10 ea) | description W × L 38 mm × 156 mm, pack of 500 ea (individually wrapped) |
| manufacturer/tradename MTC™ Bio M5431 | manufacturer/tradename MTC™ Bio M5430 | manufacturer/tradename MTC™ Bio M5421 | manufacturer/tradename MTC™ Bio M5420 |
| packaging pack of 500 ea (50 x bags 10 ea) | packaging pack of 500 ea (individually wrapped) | packaging pack of 500 ea (50 x bags 10 ea) | packaging pack of 500 ea (individually wrapped) |
| sterility sterile | sterility sterile | sterility - | sterility - |
| L × W 38 mm × 140 mm | L × W 38 mm × 140 mm | L × W - | L × W - |
Still not finding the right product?
Explore all of our products under MTC™ Bio T-Shape Spreader
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?