Przejdź do
MTCP4403
MTC™ Bio SureStand™ Multiple Pipette Stand
to hold, 3 x single pipettes OR 1 x multipipettes
Synonim(y):
pipette holder, MTC Bio P4403
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.02
UNSPSC Code:
41122801
eCl@ss:
32032209
Manufacturer/tradename:
MTC™ Bio P4403
Feature:
to hold 3 x single pipettes OR 1 x multipipettes
Material:
acrylic
Packaging:
pack of 1 ea
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
acrylic
feature
to hold 3 x single pipettes OR 1 x multipipettes
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio P4403
General description
SureStand™ Pipette Stand for 3 pipettes, up to one multi-channel, acrylic
SureStand™ Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patented “windows” allow the user to insert the multi-channel head longways through the rack while keeping the body of the pipette stable by securing the pipette at two points: under the finger hook and near the bottom of the handle. Each SureStand™ is also outfitted with four non-skid rubber “feet” for extra stability. SureStand™ is guaranteed to be the sturdiest and most versatile pipette rack in your lab.
Features and Benefits
  • Universal – Fits all popular pipettes
  • 5mm acrylic – 40% thicker
  • Non-skid rubber feet
  • Multi-channel “window” slots
  • US Patent No 872,300
Legal Information
MTC is a trademark of MTC Bio
SureStand is a trademark of MTC Bio
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
MTCP4405MTCP4408MTCP4406
description
to hold, 3 x single pipettes OR 1 x multipipettes
description
to hold, 5 x single pipettes OR 2 x multipipettes
description
to hold, 8 x single pipettes OR 6 x multipipettes
description
to hold, 6 x single pipettes OR 4 x multipipettes
feature
to hold 3 x single pipettes OR 1 x multipipettes
feature
to hold 5 x single pipettes OR 2 x multipipettes
feature
to hold 8 x single pipettes OR 6 x multipipettes
feature
to hold 6 x single pipettes OR 4 x multipipettes
manufacturer/tradename
MTC™ Bio P4403
manufacturer/tradename
MTC™ Bio P4405
manufacturer/tradename
MTC™ Bio P4408
manufacturer/tradename
MTC™ Bio P4406
material
acrylic
material
acrylic
material
acrylic
material
acrylic
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
pipette storage solution for laboratories
-
-
-
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)