Przejdź do
VM01
GenElute™ 96 Vacuum Manifold
for high-throughput purification kits
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
3310,00 zł
2813,50 zł
Informacje o tej pozycji
NACRES:
NA.52
UNSPSC Code:
41105501
eCl@ss:
32119000
2813,50 zł
Cena katalogowa3310,00 złZaoszczędź 15%
Rabat tylko przez Internet
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
General description
The GenElute 96 Vacuum Manifold is designed to provide the rapid parallel purification of nucleic acids. The manifold is capable of processing up to 96 samples simultaneously and eliminates time consuming centrifugation and decanting steps.
The GenElute Vacuum Manifold is used to support the isolation of genomic DNA, RNA and PCR products which can be used for a variety of molecular biology applications.
The GenElute 96 Vacuum Manifold provides spacer sets to accommodate suitable microtiter plates, deep well blocks (round-well and square-well blocks), or microtube racks for collecting samples.
The GenElute 96 Vacuum Manifold provides spacer sets to accommodate suitable microtiter plates, deep well blocks (round-well and square-well blocks), or microtube racks for collecting samples.
Application
The GenElute Vacuum Manifold is used to support the isolation of genomic DNA, RNA and PCR products which can be used for a variety of molecular biology applications.
Biochem/physiol Actions
The GenElute 96 Vacuum Manifold is designed to provide the rapid parallel purification of nucleic acids. The manifold is capable of processing up to 96 samples simultaneously and eliminates time consuming centrifugation and decanting steps.
Legal Information
Galacton is a trademark of Tropix, Inc.
GenElute is a trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)