Przejdź do
Z290602
Vacuum filtration assembly funnel stems
glass membrane (support), membrane diam. 47 mm, connector for 40/35 joint (fritted glass support with side arm)
Synonim(y):
Filtration Funnel Stems, Vacuum Assembly Funnel
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1940,00 zł
Informacje o tej pozycji
NACRES:
NB.24
UNSPSC Code:
41104900
Material:
glass housing, glass membrane (support)
Packaging:
pkg of 1 ea
Joint:
joint ST/NS40/35
1940,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
glass housing, glass membrane (support)
joint
joint ST/NS40/35
packaging
pkg of 1 ea
technique(s)
cell culture: suitable
membrane diam.
47 mm
fitting
connector for 40/35 joint (fritted glass support with side arm)
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z290653Z290769Z290726
description
glass membrane (support), membrane diam. 47 mm, connector for 40/35 joint (fritted glass support with side arm)
description
glass membrane (support), membrane diam. 90 mm, connector for 40/35 joint (fritted glass support with side arm)
description
stainless steel membrane (support), membrane diam. 47 mm, connector for stopper
description
glass membrane (support), membrane diam. 25 mm, connector for stopper
joint
joint ST/NS40/35
joint
joint ST/NS40/35
joint
-
joint
-
material
glass housing, glass membrane (support)
material
glass membrane (support), glass housing
material
stainless steel membrane (support), glass housing
material
glass housing, glass membrane (support)
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
fitting
connector for 40/35 joint (fritted glass support with side arm)
fitting
connector for 40/35 joint (fritted glass support with side arm)
fitting
connector for stopper
fitting
connector for stopper
membrane diam.
47 mm
membrane diam.
90 mm
membrane diam.
47 mm
membrane diam.
25 mm
technique(s)
cell culture: suitable
technique(s)
cell culture: suitable
technique(s)
cell culture: suitable
technique(s)
cell culture: suitable
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)