Przejdź do
Z290653
Vacuum filtration assembly funnel stems
glass membrane (support), membrane diam. 90 mm, connector for 40/35 joint (fritted glass support with side arm)
Synonim(y):
Vacuum Filter Assembly, Filtration Funnel, Filtration Setup
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
3690,00 zł
Informacje o tej pozycji
NACRES:
NB.24
UNSPSC Code:
41104900
Material:
glass housing, glass membrane (support)
3690,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
glass housing, glass membrane (support)
joint
joint ST/NS40/35
packaging
pkg of 1 ea
technique(s)
cell culture: suitable
membrane diam.
90 mm
fitting
connector for 40/35 joint (fritted glass support with side arm)
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z290602Z290726Z290769
description
glass membrane (support), membrane diam. 90 mm, connector for 40/35 joint (fritted glass support with side arm)
description
glass membrane (support), membrane diam. 47 mm, connector for 40/35 joint (fritted glass support with side arm)
description
glass membrane (support), membrane diam. 25 mm, connector for stopper
description
stainless steel membrane (support), membrane diam. 47 mm, connector for stopper
material
glass membrane (support), glass housing
material
glass membrane (support), glass housing
material
glass membrane (support), glass housing
material
stainless steel membrane (support), glass housing
fitting
connector for 40/35 joint (fritted glass support with side arm)
fitting
connector for 40/35 joint (fritted glass support with side arm)
fitting
connector for stopper
fitting
connector for stopper
membrane diam.
90 mm
membrane diam.
47 mm
membrane diam.
25 mm
membrane diam.
47 mm
joint
joint ST/NS40/35
joint
joint ST/NS40/35
joint
-
joint
-
technique(s)
cell culture: suitable
technique(s)
cell culture: suitable
technique(s)
cell culture: suitable
technique(s)
cell culture: suitable
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)