Przejdź do
1612743
USP
Sirolimus System Suitability Mixture
United States Pharmacopeia (USP) Reference Standard
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
UNSPSC Code:
41116107
eCl@ss:
25140904
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
grade
pharmaceutical primary standard
agency
USP
form
powder
packaging
pkg of 15 mg
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  
Other Notes
Sales restrictions may apply.
USP issued SDS can be found here
Piktogramy
Health hazard
Słowo ostrzegawcze
Warning
Kody zagrożeń
Hazard Classifications
Carc. 2 - Repr. 2
Klasa składowania
11 - Combustible Solids
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)