Przejdź do
1A05130
USP
Lamivudine Uracil Derivative
United States Pharmacopeia (USP) Reference Standard
Synonim(y):
reference material
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C8H10N2O4S
Masa cząsteczkowa:
230.24
UNSPSC Code:
41116100
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
grade
pharmaceutical analytical impurity (PAI)
agency
USP
form
powder
shelf life
at -20 °C
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-30°C
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  
Other Notes
Sales restrictions may apply.
USP issued SDS can be found here
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
135686913568581A03610
description
United States Pharmacopeia (USP) Reference Standard
description
United States Pharmacopeia (USP) Reference Standard
description
United States Pharmacopeia (USP) Reference Standard
description
Pharmaceutical Analytical Impurity (PAI)
manufacturer/tradename
USP
manufacturer/tradename
USP
manufacturer/tradename
USP
manufacturer/tradename
USP
grade
pharmaceutical analytical impurity (PAI)
grade
pharmaceutical primary standard
grade
pharmaceutical primary standard
grade
pharmaceutical analytical impurity (PAI)
application(s)
pharmaceutical small molecule
application(s)
pharmaceutical (small molecule)
pharmaceutical
application(s)
pharmaceutical (small molecule)
pharmaceutical
application(s)
pharmaceutical
pharmaceutical
pharmaceutical small molecule
shelf life
at -20 °C
shelf life
-
shelf life
-
shelf life
-
format
neat
format
mixture
format
neat
format
neat
form
powder
form
-
form
-
form
-
Klasa składowania
11 - Combustible Solids
WGK
WGK 3
Not applicable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)