Przejdź do
1A06860
USP
Orlistat Open Ring Epimer
Pharmaceutical Analytical Impurity (PAI)
Synonim(y):
reference material
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C29H55NO6
Masa cząsteczkowa:
513.75
UNSPSC Code:
41116107
eCl@ss:
25140904
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
grade
pharmaceutical analytical impurity (PAI)
agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
CCCCCCCCCCC[C@@H](C[C@H]([C@H](CCCCCC)C(=O)O)O)OC(=O)[C@H](CC(C)C)NC=O
InChI
InChI=1S/C29H55NO6/c1-5-7-9-11-12-13-14-15-16-18-24(36-29(35)26(30-22-31)20-23(3)4)21-27(32)25(28(33)34)19-17-10-8-6-2/h22-27,32H,5-21H2,1-4H3,(H,30,31)(H,33,34)/t24-,25-,26-,27+/m0/s1
InChI key
FKUNIADJSAJLGB-YIPNQBBMSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  
Other Notes
Sales restrictions may apply.
USP issued SDS can be found here
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
1A0333014788221478877
description
Pharmaceutical Analytical Impurity (PAI)
description
Pharmaceutical Analytical Impurity (PAI)
description
United States Pharmacopeia (USP) Reference Standard
description
United States Pharmacopeia (USP) Reference Standard
application(s)
pharmaceutical small molecule
application(s)
pharmaceutical small molecule
application(s)
pharmaceutical
pharmaceutical (small molecule)
application(s)
pharmaceutical
format
neat
format
neat
format
neat
format
neat
grade
pharmaceutical analytical impurity (PAI)
grade
pharmaceutical analytical impurity (PAI)
grade
pharmaceutical primary standard
grade
pharmaceutical primary standard
storage temp.
2-8°C
storage temp.
2-8°C
storage temp.
-
storage temp.
-
manufacturer/tradename
USP
manufacturer/tradename
USP
manufacturer/tradename
USP
manufacturer/tradename
USP
agency
USP
agency
USP
agency
USP
agency
USP
Piktogramy
Exclamation markEnvironment
Słowo ostrzegawcze
Warning
Kody zagrożeń
Hazard Classifications
Aquatic Acute 1 - Eye Irrit. 2
Not applicable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)