Merck
Next-gen sequencing identifies non-coding variation disrupting miRNA-binding sites in neurological disorders.
Molecular psychiatry (2017-03-16)
P Devanna, X S Chen, J Ho, D Gajewski, S D Smith, A Gialluisi, C Francks, S E Fisher, D F Newbury, S C Vernes
ABSTRAKT
Understanding the genetic factors underlying neurodevelopmental and neuropsychiatric disorders is a major challenge given their prevalence and potential severity for quality of life. While large-scale genomic screens have made major advances in this area, for many disorders the genetic underpinnings are complex and poorly understood. To date the field has focused predominantly on protein coding variation, but given the importance of tightly controlled gene expression for normal brain development and disorder, variation that affects non-coding regulatory regions of the genome is likely to play an important role in these phenotypes. Herein we show the importance of 3 prime untranslated region (3'UTR) non-coding regulatory variants across neurodevelopmental and neuropsychiatric disorders. We devised a pipeline for identifying and functionally validating putatively pathogenic variants from next generation sequencing (NGS) data. We applied this pipeline to a cohort of children with severe specific language impairment (SLI) and identified a functional, SLI-associated variant affecting gene regulation in cells and post-mortem human brain. This variant and the affected gene (ARHGEF39) represent new putative risk factors for SLI. Furthermore, we identified 3'UTR regulatory variants across autism, schizophrenia and bipolar disorder NGS cohorts demonstrating their impact on neurodevelopmental and neuropsychiatric disorders. Our findings show the importance of investigating non-coding regulatory variants when determining risk factors contributing to neurodevelopmental and neuropsychiatric disorders. In the future, integration of such regulatory variation with protein coding changes will be essential for uncovering the genetic causes of complex neurological disorders and the fundamental mechanisms underlying health and disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
Informacje o cenach i dostępności nie są obecnie dostępne.