Merck
A Label-Free Aptasensor for Ochratoxin a Detection Based on the Structure Switch of Aptamer.
Sensors (Basel, Switzerland) (2018-06-03)
Feng Liu, Ailing Ding, Jiushang Zheng, Jiucun Chen, Bin Wang
ABSTRAKT
A label-free sensing platform is developed based on switching the structure of aptamer for highly sensitive and selective fluorescence detection of ochratoxin A (OTA). OTA induces the structure of aptamer, transforms into G-quadruplex and produces strong fluorescence in the presence of zinc(II)-protoporphyrin IX probe due to the specific bind to G-quadruplex. The simple method exhibits high sensitivity towards OTA with a detection limit of 0.03 nM and excellent selectivity over other mycotoxins. In addition, the successful detection of OTA in real samples represents a promising application in food safety.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Protoporphyrin IX, ≥95%
Informacje o cenach i dostępności nie są obecnie dostępne.