Merck
Distinct Imprinting Signatures and Biased Differentiation of Human Androgenetic and Parthenogenetic Embryonic Stem Cells.
Cell stem cell (2019-09-07)
Ido Sagi, Joao C De Pinho, Michael V Zuccaro, Chen Atzmon, Tamar Golan-Lev, Ofra Yanuka, Robert Prosser, Alexandra Sadowy, Gloria Perez, Thiago Cabral, Benjamin Glaser, Stephen H Tsang, Robin Goland, Mark V Sauer, Rogerio Lobo, Nissim Benvenisty, Dieter Egli
ABSTRAKT
Genomic imprinting is an epigenetic mechanism that results in parent-of-origin monoallelic expression of specific genes, which precludes uniparental development and underlies various diseases. Here, we explored molecular and developmental aspects of imprinting in humans by generating exclusively paternal human androgenetic embryonic stem cells (aESCs) and comparing them with exclusively maternal parthenogenetic ESCs (pESCs) and bi-parental ESCs, establishing a pluripotent cell system of distinct parental backgrounds. Analyzing the transcriptomes and methylomes of human aESCs, pESCs, and bi-parental ESCs enabled the characterization of regulatory relations at known imprinted regions and uncovered imprinted gene candidates within and outside known imprinted regions. Investigating the consequences of uniparental differentiation, we showed the known paternal-genome preference for placental contribution, revealed a similar bias toward liver differentiation, and implicated the involvement of the imprinted gene IGF2 in this process. Our results demonstrate the utility of parent-specific human ESCs for dissecting the role of imprinting in human development and disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hematoxylin Solution, Gill No. 3
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
Luteinizing hormone releasing hormone, ≥97% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Iscove′s Modified Dulbecco′s Medium, liquid, sterile-filtered, With sodium bicarbonate, without L-glutamine, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
bisBenzimide H 33258, ≥98% (HPLC and TLC)