Merck
YY1 Upregulates Checkpoint Receptors and Downregulates Type I Cytokines in Exhausted, Chronically Stimulated Human T Cells.
iScience (2018-11-15)
Mumtaz Y Balkhi, Gabor Wittmann, Fang Xiong, Richard P Junghans
ABSTRAKT
T cells infiltrate affected organs in chronic infections and malignancy, but they may fail to eradicate virus-infected cells or tumor because of exhaustion. This report describes a Yin Yang-1 (YY1)-centered mechanism for diverse components that have been correlated with exhaustion. Utilizing an in vitro reconstruction of chronic T cell activation, YY1 is shown to positively regulate the checkpoint receptors PD1, Lag3, and Tim3 and to negatively regulate the type I cytokines interleukin-2 (IL-2) (in collaboration with Ezh2 histone methyltransferase) and interferon gamma (IFN-?). Other tests suggest that IL-2 failure drives a large component of cytotoxic functional decline rather than solely checkpoint receptor-ligand interactions that have been the focus of current anti-exhaustion therapies. Clinical evaluations confirm elevated YY1 and Ezh2 in melanoma tumor-infiltrating lymphocytes and in PD1+ T cells in patients with HIV. Exhaustion is revealed to be an active process as the culmination of repetitive two-signal stimulation in a feedback loop via CD3/CD28?p38MAPK/JNK?YY1? exhaustion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MEK1/2 Inhibitor III, The MEK1/2 Inhibitor III, also referenced under CAS 391210-10-9, controls the biological activity of MEK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
Przeciwciało anty-EZH2, Upstate®, from rabbit
Sigma-Aldrich
PD 98059, InSolution, ≥98%, 5 mg/ml, reversible inhibitor of MAP Kinase Kinase (MEK)
Sigma-Aldrich
SP600125, ≥98% (HPLC)